Tisk…

5-Chloro-2-hydroxybenzyl alcohol

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Chloro-2-(hydroxymethyl)phenol

OR25788-10G OR25788-1G
APOSOR25788-10GEA 6720 CZK
APOSOR25788-10G APOSOR25788-1G
5-Chloro-2-hydroxybenzyl alcohol
5-Chloro-2-hydroxybenzyl alcohol
Vzorec: C₇H₇ClO₂
MW: 158,59 g/mol
Bod tání: 84…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00021967
CAS číslo: 5330-38-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR