5-Iodo-o-toluidine
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Amino-4-iodotoluene, 5-Iodo-2-methylaniline
OR25762-25G
OR25762-5G
OR25762-100G
APOSOR25762-25GEA
953
CZK
APOSOR25762-25G
APOSOR25762-5G
APOSOR25762-100G
5-Iodo-o-toluidine
5-Iodo-o-toluidine
Vzorec:
C₇H₈IN Bod varu: 273 °C Bod tání: 48…52 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051531 CAS číslo: 83863-33-6 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...