Tisk…

3-(Cyclopentyloxy)-4-methoxybenzonitrile

Dodavatel: Apollo Scientific
Varování

OR25702-100MG OR25702-250MG OR25702-500MG
APOSOR25702-100MGEA 3940 CZK
APOSOR25702-100MG APOSOR25702-250MG APOSOR25702-500MG
3-(Cyclopentyloxy)-4-methoxybenzonitrile
3-(Cyclopentyloxy)-4-methoxybenzonitrile
Vzorec: C₁₃H₁₅NO₂
Storage Temperature: Ambient
CAS číslo: 159783-16-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR