3-Cyclopentyloxy-4-methoxybenzoic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR25693-10G
OR25693-1G
APOSOR25693-10GEA
0
CZK
APOSOR25693-10G
APOSOR25693-1G
3-Cyclopentyloxy-4-methoxybenzoic acid
3-Cyclopentyloxy-4-methoxybenzoic acid
Vzorec:
C₁₃H₁₆O₄ MW: 236,27 g/mol Bod tání: 160…164 °C Storage Temperature: Ambient |
CAS číslo:
144036-17-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...