Tisk…

Potassium methyl malonate 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: Potassium 3-methoxy-3-oxopropanoate, Monomethyl monopotassium malonate, Monomethyl malonate potassium salt

OR25586-100G OR25586-250G OR25586-500G
APOSOR25586-100GEA 992 CZK
APOSOR25586-100G APOSOR25586-250G APOSOR25586-500G
Potassium methyl malonate 98%
Potassium methyl malonate
Vzorec: C₄H₅KO₄
Bod tání: 198…205 °C
Storage Temperature: Ambient
MDL Number: MFCD00014021
CAS číslo: 38330-80-2
EINECS: 253-886-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR