2-Isopropyl-6-nitrophenol
Dodavatel: Apollo Scientific
|
Varování
|
OR25535-1G
OR25535-250MG
APOSOR25535-1GEA
11380
CZK
APOSOR25535-1G
APOSOR25535-250MG
2-Isopropyl-6-nitrophenol
2-Isopropyl-6-nitrophenol
Vzorec:
C₉H₁₁NO₃ Bod varu: 131…135 °C/20mmHg Storage Temperature: Ambient |
CAS číslo:
7545-71-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...