Tisk…

2,2-Dimethylchromane-6-carbaldehyde 85%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2,2-Dimethylchroman-6-carbaldehyde

OR25425-1G OR25425-250MG OR25425-500MG OR25425-5G
APOSOR25425-1GEA 7080 CZK
APOSOR25425-1G APOSOR25425-250MG APOSOR25425-500MG APOSOR25425-5G
2,2-Dimethylchromane-6-carbaldehyde 85%
2,2-Dimethylchromane-6-carbaldehyde
Vzorec: C₁₂H₁₄O₂
MW: 190,24 g/mol
Bod varu: 110…114 °C/2mmHg
Storage Temperature: Ambient
MDL Number: MFCD00052286
CAS číslo: 61370-75-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR