Tisk…

Ethyl-4-bromo-1,3-dimethylpyrazole-5-carboxylate

Dodavatel: Apollo Scientific
Varování

OR2540-1G OR2540-250MG OR2540-5G
APOSOR2540-1GEA 3970 CZK
APOSOR2540-1G APOSOR2540-250MG APOSOR2540-5G
Ethyl-4-bromo-1,3-dimethylpyrazole-5-carboxylate
Ethyl-4-bromo-1,3-dimethylpyrazole-5-carboxylate
Vzorec: C₈H₁₁BrN₂O₂
MW: 247,1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02253803
CAS číslo: 5775-89-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR