Tisk…

2-Amino-4-(4-pyridyl)thiazole

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-(Pyridin-4-yl)thiazol-2-amine

OR2532-1G OR2532-5G
APOSOR2532-1GEA 1330 CZK
APOSOR2532-1G APOSOR2532-5G
2-Amino-4-(4-pyridyl)thiazole
2-Amino-4-(4-pyridyl)thiazole
Vzorec: C₈H₇N₃S
MW: 177,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00490009
CAS číslo: 30235-28-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR