Tisk…

Piperidine-1-carboximidamide hydroiodide

Dodavatel: Apollo Scientific

OR25055-1G OR25055-250MG OR25055-500MG OR25055-2.5G OR25055-5G
APOSOR25055-1GEA 4860 CZK
APOSOR25055-1G APOSOR25055-250MG APOSOR25055-500MG APOSOR25055-2.5G APOSOR25055-5G
Piperidine-1-carboximidamide hydroiodide
Piperidine-1-carboximidamide hydroiodide
Vzorec: C₆H₁₄IN₃
Storage Temperature: Ambient
MDL Number: MFCD00114187
CAS číslo: 102392-91-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR