(4-Chlorophenylthio)acetonitrile
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
[(4-Chlorophenyl)sulphanyl]acetonitrile
OR25038-1G
OR25038-5G
OR25038-25G
APOSOR25038-1GEA
622
CZK
APOSOR25038-1G
APOSOR25038-5G
APOSOR25038-25G
(4-Chlorophenylthio)acetonitrile
(4-Chlorophenylthio)acetonitrile
Vzorec:
C₈H₆ClNS MW: 183,66 g/mol Bod tání: 84…90 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00019832 CAS číslo: 18527-19-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...