2,6-Pyridinedicarboxaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
OR24830-1G
OR24830-5G
OR24830-25G
APOSOR24830-1GEA
1120
CZK
APOSOR24830-1G
APOSOR24830-5G
APOSOR24830-25G
2,6-Pyridinedicarboxaldehyde
2,6-Pyridinedicarboxaldehyde
Vzorec:
C₇H₅NO₂ MW: 135.12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00010103 CAS číslo: 5431-44-7 EINECS: 226-589-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...