Azetidin-2-one 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Azetidinone
OR24812-5G
OR24812-1G
APOSOR24812-5GEA
3540
CZK
APOSOR24812-5G
APOSOR24812-1G
Azetidin-2-one 98%
Azetidin-2-one
Vzorec:
C₃H₅NO MW: 71,08 g/mol Bod varu: 106 °C/15mmHg Bod tání: 72…77 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00013328 CAS číslo: 930-21-2 UN: 3263 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...