Methyl hydrazinecarbohydrazonothioate hydroiodide
Dodavatel: Apollo Scientific
|
Varování
|
OR24599-1G
OR24599-5G
APOSOR24599-1GEA
1120
CZK
APOSOR24599-1G
APOSOR24599-5G
Methyl hydrazinecarbohydrazonothioate hydroiodide
Methyl hydrazinecarbohydrazonothioate hydroiodide
Vzorec:
C₂H₉IN₄S MW: 248,09 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09991850 CAS číslo: 37839-01-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...