Tisk…

N-(tert-Butyl)hydroxylamine hydrochloride 95+%

Dodavatel: Apollo Scientific
Varování

Synonyms: N-(tert-Butyl) hydroxylamine hydrochloride

OR24575-1G OR24575-5G OR24575-10G
APOSOR24575-1GEA 662 CZK
APOSOR24575-1G APOSOR24575-5G APOSOR24575-10G
N-(tert-Butyl)hydroxylamine hydrochloride 95+%
N-(tert-Butyl)hydroxylamine hydrochloride
Vzorec: C₄H₁₁NO
MW: 125.60 g/mol
Bod tání: 183…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00012598
CAS číslo: 57497-39-9
EINECS: 260-771-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR