2,3-Dihydro-5-(2-hydroxyethyl)benzo[b]furan
Dodavatel: Apollo Scientific
|
Varování
|
OR2452-1G
OR2452-250MG
APOSOR2452-1GEA
21490
CZK
APOSOR2452-1G
APOSOR2452-250MG
2,3-Dihydro-5-(2-hydroxyethyl)benzo[b]furan
2,3-Dihydro-5-(2-hydroxyethyl)benzo[b]furan
Vzorec:
C₁₀H₁₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD06797643 CAS číslo: 87776-76-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...