Tisk…

Methyl-3-isothiocyanato-4-methylthiophene-2-carboxylate

Dodavatel: Apollo Scientific
Varování

OR24321-1G OR24321-5G OR24321-10G
APOSOR24321-1GEA 1240 CZK
APOSOR24321-1G APOSOR24321-5G APOSOR24321-10G
Methyl-3-isothiocyanato-4-methylthiophene-2-carboxylate
Methyl-3-isothiocyanato-4-methylthiophene-2-carboxylate
Vzorec: C₈H₇NO₂S₂
MW: 213,28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00052073
CAS číslo: 81321-15-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR