Tisk…

2,4-Dimethylbenzoyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2,4-Dimethylbenzoylchloride, 2,4-Dimethylbenzene-1-carbonyl chloride

OR24121-250MG OR24121-100MG
APOSOR24121-250MGEA 705 CZK
APOSOR24121-250MG APOSOR24121-100MG
2,4-Dimethylbenzoyl chloride
2,4-Dimethylbenzoyl chloride
Vzorec: C₉H₉ClO
MW: 168.62
Bod varu: 82 °C/1mmHg
Bod tání: 24…26 °C
Storage Temperature: Ambient
MDL Number: MFCD00798005
CAS číslo: 21900-42-5
UN: 3265
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR