syn-Pyridine-2-carboxaldoxime
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
syn-2-Pyridinealdoxime
OR24107-100G
OR24107-25G
OR24107-5G
APOSOR24107-100GEA
3540
CZK
APOSOR24107-100G
APOSOR24107-25G
APOSOR24107-5G
syn-Pyridine-2-carboxaldoxime
syn-Pyridine-2-carboxaldoxime
Vzorec:
C₆H₆N₂O MW: 122,13 g/mol Bod tání: 110…112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00167166 CAS číslo: 1193-96-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...