Tisk…

Chitin

Dodavatel: Apollo Scientific
Varování

OR2403-250G OR2403-25G
APOSOR2403-250GEA 3810 CZK
APOSOR2403-250G APOSOR2403-25G
Chitin
Chitin
Vzorec: C₃₀H₅₀N₄O₁₉
Storage Temperature: Ambient
MDL Number: MFCD00466914
CAS číslo: 1398-61-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR