Tisk…

2,5-Dichloro-4-nitro-3-thiophenesulphonyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

OR23993-1G OR23993-250MG OR23993-5G
APOSOR23993-1GEA 6580 CZK
APOSOR23993-1G APOSOR23993-250MG APOSOR23993-5G
2,5-Dichloro-4-nitro-3-thiophenesulphonyl chloride
2,5-Dichloro-4-nitro-3-thiophenesulphonyl chloride
Vzorec: C₄Cl₃NO₄S₂
Storage Temperature: Ambient
MDL Number: MFCD01570024
CAS číslo: 59768-12-6
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR