Tisk…

5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulphonyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

OR23902-1G OR23902-250MG OR23902-5G
APOSOR23902-1GEA 744 CZK
APOSOR23902-1G APOSOR23902-250MG APOSOR23902-5G
5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulphonyl chloride
5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulphonyl chloride
Vzorec: C₅H₆Cl₂N₂O₂S
Bod varu: 105 °C/1mmHg
Storage Temperature: Ambient
MDL Number: MFCD00052536
CAS číslo: 88398-93-0
UN: 3261
ADR: 8,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR