Tisk…

Ethyl 2-((dimethylamino)methylene)-3-oxobutanoate

Dodavatel: Apollo Scientific
Varování

Synonyms: Ethyl 2-((dimethylamino)methylene)-3-oxobutanoate, Ethyl 2-acetyl-3-(dimethylamino)acrylate

OR23897-1G OR23897-5G
APOSOR23897-1GEA 622 CZK
APOSOR23897-1G APOSOR23897-5G
Ethyl 2-((dimethylamino)methylene)-3-oxobutanoate
Ethyl-2-((dimethylamino)methylene)-3-oxobutanoate
Vzorec: C₉H₁₅NO₃
Storage Temperature: Ambient
MDL Number: MFCD00087563
CAS číslo: 51145-57-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR