Tisk…

ω-Bromo-2-nitroacetophenone

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: α-Bromo-2-nitroacetophenone, 2-Bromo-2'-nitroacetophenone, Bromomethyl o-nitrophenyl ketone, 2-Nitrophenacyl bromide

OR2385-1G OR2385-25G OR2385-5G
APOSOR2385-1GEA 622 CZK
APOSOR2385-1G APOSOR2385-25G APOSOR2385-5G
ω-Bromo-2-nitroacetophenone
ω-Bromo-2-nitroacetophenone
Vzorec: C₈H₆BrNO₃
MW: 244,04 g/mol
Bod tání: 53…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00010294
CAS číslo: 6851-99-6
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR