Tisk…

tert-Butoxybis(dimethylamino)methane 90%

Dodavatel: Apollo Scientific
Varování

Synonyms: Bredereck's reagent, 1-tert-Butoxy-N,N,N',N'-tetramethylmethylenenediamine, [(tert-butoxy)(dimethylamino)methyl]dimethylamine, tert-Butoxy bis(dimethylamino)methane

OR2361-100G OR2361-25G
APOSOR2361-100GEA 2980 CZK
APOSOR2361-100G APOSOR2361-25G
tert-Butoxybis(dimethylamino)methane 90%
tert-Butoxybis(dimethylamino)methane
Vzorec: C₉H₂₂N₂O
MW: 174,288 g/mol
Bod varu: 50…55 °C/15mmHg
Hustota: 0,844 g/cm³ (25 °C)
Bod vzplanutí: 41 °C
Storage Temperature: Freezer
MDL Number: MFCD00042858
CAS číslo: 5815-08-7
EINECS: 227-383-9
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR