Tisk…

Mucic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: Galactaric acid, rel-(2R,3S,4R,5S)-2,3,4,5-Tetrahydroxyhexanedioic acid, (2R,3S,4R,5S)-2,3,4,5-Tetrahydroxyhexanedioic acid

OR2356-100G OR2356-1KG OR2356-500G
APOSOR2356-100GEA 622 CZK
APOSOR2356-100G APOSOR2356-1KG APOSOR2356-500G
Mucic acid 97%
Mucic acid
Vzorec: C₆H₁₀O₈
MW: 210,14 g/mol
Bod tání: 210…230 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00004239
CAS číslo: 526-99-8
EINECS: 208-404-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR