2',3',4',5',6'-Pentamethyl-2-phenylacetophenone
Dodavatel: Apollo Scientific
Synonyms:
1-(2,3,4,5,6-Pentamethylphenyl)-2-phenylethan-1-one
OR23525-10G
OR23525-1G
OR23525-5G
APOSOR23525-10GEA
9860
CZK
APOSOR23525-10G
APOSOR23525-1G
APOSOR23525-5G
2',3',4',5',6'-Pentamethyl-2-phenylacetophenone
2',3',4',5',6'-Pentamethyl-2-phenylacetophenone
Vzorec:
C₁₉H₂₂O MW: 266,38 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00052403 CAS číslo: 147834-57-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...