2-(2,3,4,5,6-Pentamethylbenzoyl)benzoic acid
Dodavatel: Apollo Scientific
OR23506-10G
OR23506-1G
OR23506-5G
APOSOR23506-10GEA
8440
CZK
APOSOR23506-10G
APOSOR23506-1G
APOSOR23506-5G
2-(2,3,4,5,6-Pentamethylbenzoyl)benzoic acid
2-(2,3,4,5,6-Pentamethylbenzoyl)benzoic acid
Vzorec:
C₁₉H₂₀O₃ MW: 296,37 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00051629 CAS číslo: 111385-66-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...