Tisk…

2-Chloro-2',3',4',5',6'-pentamethylacetophenone

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2,3,4,5,6-Pentamethylphenacyl chloride

OR23406-1G OR23406-250MG
APOSOR23406-1GEA 5290 CZK
APOSOR23406-1G APOSOR23406-250MG
2-Chloro-2',3',4',5',6'-pentamethylacetophenone
2-Chloro-2',3',4',5',6'-pentamethylacetophenone
Vzorec: C₁₃H₁₇ClO
Bod tání: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00117891
CAS číslo: 57196-63-1
UN: 3261
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR