Tisk…

4-(1H-Pyrrol-1-yl)benzylamine

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: [4-(1H-Pyrrol-1-yl)phenyl]methylamine

OR23401-1G OR23401-250MG
APOSOR23401-1GEA 13790 CZK
APOSOR23401-1G APOSOR23401-250MG
4-(1H-Pyrrol-1-yl)benzylamine
4-(1H-Pyrrol-1-yl)benzylamine
Vzorec: C₁₁H₁₂N₂
MW: 172,23 g/mol
Bod tání: 81…84 °C
Storage Temperature: Ambient
MDL Number: MFCD03659728
CAS číslo: 465514-27-6
UN: 3259
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR