Tisk…

4-(4-Methylpiperazin-1-yl)benzylamine 98%

Dodavatel: Apollo Scientific
Varování

OR23311-25G OR23311-5G
APOSOR23311-25GEA 17360 CZK
APOSOR23311-25G APOSOR23311-5G
4-(4-Methylpiperazin-1-yl)benzylamine 98%
4-(4-Methylpiperazin-1-yl)benzylamine
Vzorec: C₁₂H₁₉N₃
Bod varu: 122…124 °C/0,05mmHg
Storage Temperature: Ambient
MDL Number: MFCD03086117
CAS číslo: 216144-45-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR