3-(Chloromethyl)benzoic acid
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR2331-1G
OR2331-25G
OR2331-5G
APOSOR2331-1GEA
622
CZK
APOSOR2331-1G
APOSOR2331-25G
APOSOR2331-5G
3-(Chloromethyl)benzoic acid
3-(Chloromethyl)benzoic acid
Vzorec:
C₈H₇ClO₂ Bod tání: 136…138 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191922 CAS číslo: 31719-77-4 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...