4-(1H-Imidazol-1-yl)benzoic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-(1-Imidazolyl)benzoic acid
OR23298-1G
OR23298-500MG
OR23298-5G
APOSOR23298-1GEA
1210
CZK
APOSOR23298-1G
APOSOR23298-500MG
APOSOR23298-5G
4-(1H-Imidazol-1-yl)benzoic acid
4-(1H-Imidazol-1-yl)benzoic acid
Vzorec:
C₁₀H₈N₂O₂ MW: 188,19 g/mol Bod tání: 305 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00586645 CAS číslo: 17616-04-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...