4-(1H-Pyrazol-1-yl)benzoic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR23288-1G
OR23288-250MG
OR23288-5G
APOSOR23288-1GEA
705
CZK
APOSOR23288-1G
APOSOR23288-250MG
APOSOR23288-5G
4-(1H-Pyrazol-1-yl)benzoic acid
4-(1H-Pyrazol-1-yl)benzoic acid
Vzorec:
C₁₀H₈N₂O₂ MW: 188,18 g/mol Bod tání: 263…272 °C Storage Temperature: Ambient |
MDL Number:
MFCD02682056 CAS číslo: 16209-00-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...