(±)-Tetrahydrofurfuryl bromide 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR23265-100G
OR23265-500G
APOSOR23265-100GEA
4630
CZK
APOSOR23265-100G
APOSOR23265-500G
(±)-Tetrahydrofurfuryl bromide 95%
(±)-Tetrahydrofurfuryl bromide
Vzorec:
C₅H₉BrO Bod varu: 170…171 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00016894 CAS číslo: 1192-30-9 EINECS: 214-750-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...