3-Thienyl isocyanate
Dodavatel: Apollo Scientific
|
Varování
|
OR23202-250MG
APOSOR23202-250MGEA
3380
CZK
APOSOR23202-250MG
3-Thienyl isocyanate
3-Thienyl isocyanate
- May form small amount of sediment.
Vzorec:
C₅H₃NOS MW: 125,15 g/mol Bod varu: 36…37 °C/0,75mmHg Storage Temperature: Refrigerator |
MDL Number:
MFCD02681899 CAS číslo: 76536-95-3 UN: 2206 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...