Tisk…

3-Thiophenecarbonyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

OR23199-1G OR23199-5G
APOSOR23199-1GEA 0 CZK
APOSOR23199-1G APOSOR23199-5G
3-Thiophenecarbonyl chloride
3-Thiophenecarbonyl chloride
Vzorec: C₅H₃ClOS
MW: 146,588 g/mol
Bod tání: 49…51 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00130090
CAS číslo: 41507-35-1
EINECS: 255-420-9
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR