Thiophene-2-carbonyl chloride 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Thiophenecarbonyl chloride, Thiophene-2-carboxylic acid chloride
OR23195-100G
OR23195-500G
APOSOR23195-100GEA
1240
CZK
APOSOR23195-100G
APOSOR23195-500G
Thiophene-2-carbonyl chloride 98%
Thiophene-2-carbonyl chloride
Vzorec:
C₅H₃ClOS MW: 146,6 g/mol Bod varu: 206…208 °C Hustota: 1,371 g/cm³ (25 °C) Bod vzplanutí: 91 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005428 CAS číslo: 5271-67-0 EINECS: 226-092-4 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...