1-(1-Benzothiophen-3-yl)-2-bromo-1-ethanone 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
3-(Bromoacetyl)benzo[b]thiophene
OR23175-1G
OR23175-250MG
APOSOR23175-1GEA
2280
CZK
APOSOR23175-1G
APOSOR23175-250MG
1-(1-Benzothiophen-3-yl)-2-bromo-1-ethanone 97%
1-(1-Benzothiophen-3-yl)-2-bromo-1-ethanone
Vzorec:
C₁₀H₇BrOS MW: 255,14 g/mol Bod tání: 138…139 °C Storage Temperature: Ambient |
MDL Number:
MFCD00176410 CAS číslo: 26167-45-3 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...