Benzo[b]thiophene-3-carbonyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR23172-1G
OR23172-5G
APOSOR23172-1GEA
0
CZK
APOSOR23172-1G
APOSOR23172-5G
Benzo[b]thiophene-3-carbonyl chloride
Benzo[b]thiophene-3-carbonyl chloride
Vzorec:
C₉H₅ClOS Bod varu: 122…124 °C/1mmHg Storage Temperature: Ambient |
CAS číslo:
39827-12-8 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...