Tisk…

Benzo[b]thiophene-3-carboxylic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Benzothiophene-3-carboxylic acid

OR23171-1G OR23171-250MG OR23171-5G
APOSOR23171-1GEA 2820 CZK
APOSOR23171-1G APOSOR23171-250MG APOSOR23171-5G
Benzo[b]thiophene-3-carboxylic acid
Benzo[b]thiophene-3-carboxylic acid
Vzorec: C₉H₆O₂S
MW: 178,22 g/mol
Bod tání: 174…176 °C
Storage Temperature: Ambient
MDL Number: MFCD01846406
CAS číslo: 5381-25-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR