4-Thien-2-ylbenzoyl chloride
Dodavatel: Apollo Scientific
OR23164-1G
OR23164-250MG
OR23164-500MG
APOSOR23164-1GEA
4070
CZK
APOSOR23164-1G
APOSOR23164-250MG
APOSOR23164-500MG
4-Thien-2-ylbenzoyl chloride
4-Thien-2-ylbenzoyl chloride
Vzorec:
C₁₁H₇ClOS Storage Temperature: Ambient |
MDL Number:
MFCD02681905 CAS číslo: 181132-70-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...