5-(Thiophen-2-yl)nicotinic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR23158-1G
OR23158-250MG
APOSOR23158-1GEA
6220
CZK
APOSOR23158-1G
APOSOR23158-250MG
5-(Thiophen-2-yl)nicotinic acid 97%
5-(Thiophen-2-yl)nicotinic acid
Vzorec:
C₁₀H₇NO₂S MW: 205,24 g/mol Bod tání: 260…261 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02682027 CAS číslo: 306934-96-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...