5-Phenylthiophene-2-carboxylic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Phenyl-2-thiophenecarboxylic acid, 5-Phenyl-2-thenoic acid
OR23146-1G
OR23146-250MG
OR23146-5G
APOSOR23146-1GEA
3110
CZK
APOSOR23146-1G
APOSOR23146-250MG
APOSOR23146-5G
5-Phenylthiophene-2-carboxylic acid 97%
5-Phenylthiophene-2-carboxylic acid
Vzorec:
C11H8O2S MW: 204,2450 g/mol Bod tání: 188…192 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01313432 CAS číslo: 19163-24-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...