2,1,3-Benzoxadiazole-5-carbaldehyde 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Benzo[c][1,2,5]oxadiazole-5-carbaldehyde
OR23134-1G
OR23134-250MG
APOSOR23134-1GEA
3480
CZK
APOSOR23134-1G
APOSOR23134-250MG
2,1,3-Benzoxadiazole-5-carbaldehyde 97%
2,1,3-Benzoxadiazole-5-carbaldehyde
Vzorec:
C₇H₄N₂O₂ MW: 148,12 g/mol Bod tání: 58 °C Storage Temperature: Ambient |
MDL Number:
MFCD01200350 CAS číslo: 32863-33-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...