2,1,3-Benzothiadiazol-5-ylamine 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Amino-2,1,3-benzothiadiazole, Benzo[c][1,2,5]thiadiazol-5-amine
OR23132-10G
OR23132-1G
OR23132-5G
APOSOR23132-10GEA
14780
CZK
APOSOR23132-10G
APOSOR23132-1G
APOSOR23132-5G
2,1,3-Benzothiadiazol-5-ylamine 97%
2,1,3-Benzothiadiazol-5-ylamine
Vzorec:
C₆H₅N₃S Bod tání: 112…115 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00185826 CAS číslo: 874-37-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...