Tisk…

1H-Benzimidazole-2-carboxylic acid

Dodavatel: Apollo Scientific
Nebezpečí

OR23116-1G OR23116-25G OR23116-5G
APOSOR23116-1GEA 622 CZK
APOSOR23116-1G APOSOR23116-25G APOSOR23116-5G
1H-Benzimidazole-2-carboxylic acid
1H-Benzimidazole-2-carboxylic acid
Vzorec: C₈H₆N₂O₂
MW: 162,1500 g/mol
Bod tání: 176…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00721565
CAS číslo: 2849-93-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR