Tisk…

2-Amino-1,3,4-thiadiazole

Dodavatel: Apollo Scientific
Nebezpečí

OR23108-100G OR23108-25G OR23108-5G
APOSOR23108-100GEA 827 CZK
APOSOR23108-100G APOSOR23108-25G APOSOR23108-5G
2-Amino-1,3,4-thiadiazole
2-Amino-1,3,4-thiadiazole
Vzorec: C₂H₃N₃S
Storage Temperature: Ambient
MDL Number: MFCD00003107
CAS číslo: 4005-51-0
EINECS: 223-657-7
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR