Tisk…

Benzo[b]furan-2-boronic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: Benzofuran-2-boronic acid, 2-Benzofurylboronic acid

OR23097-5G OR23097-25G OR23097-100G
APOSOR23097-5GEA 1660 CZK
APOSOR23097-5G APOSOR23097-25G APOSOR23097-100G
Benzo[b]furan-2-boronic acid 98%
Benzo[b]furan-2-boronic acid
Vzorec: C₈H₇BO₃
MW: 161,95 g/mol
Bod tání: 196…198 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00236019
CAS číslo: 98437-24-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR