2,4-Dimethyl-1,3-thiazole-5-carbaldehyde
Dodavatel: Apollo Scientific
Synonyms:
2,4-Dimethyl-1,3-thiazole-5-carboxaldehyde
OR23082-1G
OR23082-50MG
OR23082-100MG
OR23082-250MG
OR23082-500MG
OR23082-2.5G
APOSOR23082-1GEA
3380
CZK
APOSOR23082-1G
APOSOR23082-50MG
APOSOR23082-100MG
APOSOR23082-250MG
APOSOR23082-500MG
APOSOR23082-2.5G
2,4-Dimethyl-1,3-thiazole-5-carbaldehyde
2,4-Dimethyl-1,3-thiazole-5-carbaldehyde
Vzorec:
C₆H₇NOS Bod tání: 23…27 °C Bod vzplanutí: 98 °C Storage Temperature: Ambient |
MDL Number:
MFCD03407320 CAS číslo: 95453-54-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...